Assay Details
Gene Symbol: LOC100911615 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749014.3 | XP_003749062.1 | 1 - 2 | 317 | 115 |
Gene Symbol: Pnpla2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108509.2 | NP_001101979.2 | 306 | 115 | ||
GenBank mRNA | CV120368.1 | - | 1 - 2 | 339 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
