Assay Details
Gene Symbol: Mamdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760316.1 | XP_008758538.1 | 8 - 9 | 1827 | 121 | |
GenBank mRNA | BC079474.1 | - | 2 - 3 | 181 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
