Assay Details
Gene Symbol: Mamdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760316.1 | XP_008758538.1 | 6 - 7 | 1462 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
