Assay Details
Gene Symbol: Bcl2l12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108480.1 | NP_001101950.1 | 6 - 7 | 708 | 65 | |
XM_006229097.2 | XP_006229159.1 | 5 - 6 | 681 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
