Assay Details
Gene Symbol: Tomm40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212520.1 | NP_997685.1 | 2 - 3 | 342 | 85 | |
XM_006228420.2 | XP_006228482.1 | 3 - 4 | 450 | 85 | ||
GenBank mRNA | AB162855.1 | - | 2 - 3 | 342 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
