Assay Details
Gene Symbol: Bax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017059.2 | NP_058755.2 | 2 - 3 | 182 | 76 | |
GenBank mRNA | AF235993.1 | - | 1 - 2 | 547 | 76 | |
CB586216.1 | - | 2 - 3 | 182 | 76 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF235993.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
