Assay Details
Gene Symbol: Gtf2h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108485.1 | NP_001101955.1 | 8 - 9 | 1163 | 142 | |
XM_006229235.1 | XP_006229297.1 | 8 - 9 | 1434 | 142 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
