Assay Details
Gene Symbol: Gtf2h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108485.1 | NP_001101955.1 | 12 - 13 | 1548 | 91 | |
XM_006229235.1 | XP_006229297.1 | 12 - 13 | 1819 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
nucleotide-excision repair
transcription by RNA polymerase II
protein phosphorylation
positive regulation of transcription by RNA polymerase II
chromatin binding
protein kinase activity
protein binding
ATP-dependent activity, acting on DNA
RNA polymerase II CTD heptapeptide repeat kinase activity
nucleotide-excision repair
transcription by RNA polymerase II
protein phosphorylation
positive regulation of transcription by RNA polymerase II
chromatin binding
protein kinase activity
protein binding
ATP-dependent activity, acting on DNA
RNA polymerase II CTD heptapeptide repeat kinase activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P461085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107544 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513575 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580996 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210843 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665301 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309077 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964845 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688201 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887118 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127960 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34241 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846411 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505584 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701494 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596454 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543875 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736139 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46542 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52993 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152912 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P461085 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P461085 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P461085 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P492053 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P461085 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 6460 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10707130 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10707129 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10707134 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10707133 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10707124 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10707128 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10707131 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10707127 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10707125 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10707123 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10707136 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10707122 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10707132 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10707126 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10707121 | - | - |
Rat Genome 230 2.0 Array | 1397557_PM_at | - | - |
Rat Genome 230 2.0 Array | 1397557_at | - | - |
Rat Genome 230 2.0 Array | 1373094_PM_at | - | - |
Rat Genome 230 2.0 Array | 1373094_at | - | - |
Affymetrix HT Rat Focus Array | 1397557_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1373094_at | - | - |
Affymetrix HT Rat Focus Array | 1373094_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1397557_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100012348.rn.1 | TC0100001527.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100012347.rn.1 | TC0100001527.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100012345.rn.1 | TC0100001527.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100012346.rn.1 | TC0100001527.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100012343.rn.1 | TC0100001527.rn.1 | RTA TC |
Rat Expression Set 230 | 1373094_PM_at | - | - |
Rat Expression Set 230 | 1373094_at | - | - |
Rat Expression Set 230 | 1397557_at | - | - |
Rat Expression Set 230 | 1397557_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10707121 | - | - |
Rat Genome U34 Set | rc_AI059759_at | - | - |
Rat Genome U34 Set | rc_AA849847_at | - | - |
Rat Genome U34 Set | rc_AI229307_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357684 | - | - |
