Assay Details
Gene Symbol: Psmc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057122.1 | NP_476463.1 | 3 - 4 | 333 | 67 | |
GenBank mRNA | BC063145.1 | - | 3 - 4 | 343 | 6889167 | 67 |
D50695.1 | - | 3 - 4 | 333 | 67 | ||
FQ225557.1 | - | 3 - 4 | 358 | 67 | ||
FQ226113.1 | - | 3 - 4 | 342 | 67 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
