Assay Details
Gene Symbol: Napa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080585.1 | NP_542152.1 | 9 - 10 | 781 | 79 | |
GenBank mRNA | BC063156.1 | - | 9 - 10 | 840 | 79 | |
FQ213217.1 | - | 9 - 10 | 785 | 79 | ||
FQ216375.1 | - | 9 - 10 | 1099 | 79 | ||
FQ234471.1 | - | 3 - 4 | 244 | 79 | ||
X89968.1 | - | 9 - 10 | 781 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
