Assay Details
Gene Symbol: Suds3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769301.1 | XP_008767523.1 | 6 - 7 | 657 | 88 | |
XM_341092.7 | XP_341093.4 | 6 - 7 | 656 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
