Assay Details
Gene Symbol: Suds3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_341092.7 | XP_341093.4 | 9 - 10 | 836 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
