Assay Details
Gene Symbol: Eif2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172029.2 | NP_742026.1 | 5 - 6 | 522 | 140 | |
GenBank mRNA | BC081709.1 | - | 5 - 6 | 522 | 140 | |
FQ217438.1 | - | 5 - 6 | 560 | 140 | ||
U05821.1 | - | 5 - 6 | 541 | 140 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
