Assay Details
Gene Symbol: Eif2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172029.2 | NP_742026.1 | 8 - 9 | 797 | 138 | |
GenBank mRNA | BC081709.1 | - | 8 - 9 | 797 | 7135576 | 138 |
U05821.1 | - | 8 - 9 | 816 | 138 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
