Assay Details
Gene Symbol: Oas1k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009489.1 | NP_001009489.1 | 1132 | 67 | ||
GenBank mRNA | AY196696.1 | - | 1132 | 67 | ||
BC088437.1 | - | 1081 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
