Assay Details
Gene Symbol: Diablo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008292.1 | NP_001008293.1 | 1 - 2 | 70 | 76 | |
GenBank mRNA | BC086366.1 | - | 1 - 2 | 70 | 7305418 | 76 |
FQ219826.1 | - | 1 - 2 | 63 | 76 |
Gene Symbol: LOC100360940 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002729540.3 | XP_002729586.2 | 1 | 100 | 76 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
