Assay Details
Gene Symbol: Aldh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032416.1 | NP_115792.1 | 10 - 11 | 1284 | 95 | |
XM_006249385.2 | XP_006249447.1 | 10 - 11 | 1348 | 95 | ||
GenBank mRNA | AF529165.2 | - | 10 - 11 | 1161 | 95 | |
AY566467.1 | - | 10 - 11 | 1228 | 95 | ||
AY566468.1 | - | 10 - 11 | 1228 | 95 | ||
AY566469.1 | - | 10 - 11 | 1228 | 95 | ||
BC062081.1 | - | 10 - 11 | 1260 | 95 | ||
FQ210970.1 | - | 2 - 3 | 299 | 95 | ||
X14977.1 | - | 10 - 11 | 1284 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
