Assay Details
Gene Symbol: Aldh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032416.1 | NP_115792.1 | 8 - 9 | 940 | 64 | |
XM_006249385.2 | XP_006249447.1 | 8 - 9 | 1004 | 64 | ||
GenBank mRNA | AF529165.2 | - | 8 - 9 | 817 | 64 | |
AY566467.1 | - | 8 - 9 | 884 | 64 | ||
AY566468.1 | - | 8 - 9 | 884 | 64 | ||
AY566469.1 | - | 8 - 9 | 884 | 64 | ||
BC062081.1 | - | 8 - 9 | 916 | 5621229 | 64 | |
X14977.1 | - | 8 - 9 | 940 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
