Assay Details
Gene Symbol: P2rx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019256.1 | NP_062129.1 | 12 - 13 | 1408 | 119 | |
XM_008769223.1 | XP_008767445.1 | 12 - 13 | 1582 | 119 | ||
GenBank mRNA | FJ436445.1 | - | 12 - 13 | 1446 | 119 | |
X95882.1 | - | 12 - 13 | 1408 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
cell morphogenesis
positive regulation of cytokine production
phagolysosome assembly
positive regulation of T cell mediated cytotoxicity
positive regulation of protein phosphorylation
regulation of sodium ion transport
protein phosphorylation
membrane protein ectodomain proteolysis
phospholipid transfer to membrane
cation transport
calcium ion transport
cell volume homeostasis
vesicle budding from membrane
inflammatory response
mitochondrion organization
plasma membrane organization
cell surface receptor signaling pathway
response to xenobiotic stimulus
response to mechanical stimulus
response to bacterium
response to organic substance
response to zinc ion
gene expression
positive regulation of calcium ion transport into cytosol
programmed cell death
positive regulation of glutamate secretion
positive regulation of gamma-aminobutyric acid secretion
response to organic cyclic compound
organic cation transport
synaptic vesicle exocytosis
protein processing
neuronal action potential
sensory perception of pain
cytolysis
protein catabolic process
positive regulation of bone mineralization
cellular response to extracellular stimulus
bleb assembly
positive regulation of prostaglandin secretion
response to lipopolysaccharide
positive regulation of interleukin-1 beta production
positive regulation of interleukin-6 production
collagen metabolic process
response to ATP
response to fluid shear stress
purinergic nucleotide receptor signaling pathway
T cell proliferation
T cell homeostasis
positive regulation of apoptotic process
positive regulation of catalytic activity
NAD transport
negative regulation of MAPK cascade
positive regulation of MAPK cascade
phospholipid translocation
positive regulation of ossification
negative regulation of bone resorption
positive regulation of cytolysis
ceramide biosynthetic process
pore complex assembly
skeletal system morphogenesis
homeostasis of number of cells within a tissue
positive regulation of protein secretion
defense response to Gram-positive bacterium
release of sequestered calcium ion into cytosol
protein complex oligomerization
positive regulation of cytoskeleton organization
response to calcium ion
response to electrical stimulus
membrane depolarization
positive regulation of mitochondrial depolarization
positive regulation of lymphocyte apoptotic process
cellular response to organic cyclic compound
reactive oxygen species metabolic process
apoptotic signaling pathway
extrinsic apoptotic signaling pathway
cation transmembrane transport
magnesium ion binding
lipopolysaccharide binding
purinergic nucleotide receptor activity
extracellularly ATP-gated cation channel activity
signaling receptor binding
copper ion binding
protein binding
ATP binding
zinc ion binding
channel activity
signaling receptor activity
protein serine/threonine kinase activator activity
scaffold protein binding
cell morphogenesis
positive regulation of cytokine production
phagolysosome assembly
positive regulation of T cell mediated cytotoxicity
positive regulation of protein phosphorylation
regulation of sodium ion transport
protein phosphorylation
membrane protein ectodomain proteolysis
phospholipid transfer to membrane
cation transport
calcium ion transport
cell volume homeostasis
vesicle budding from membrane
inflammatory response
mitochondrion organization
plasma membrane organization
cell surface receptor signaling pathway
response to xenobiotic stimulus
response to mechanical stimulus
response to bacterium
response to organic substance
response to zinc ion
gene expression
positive regulation of calcium ion transport into cytosol
programmed cell death
positive regulation of glutamate secretion
positive regulation of gamma-aminobutyric acid secretion
response to organic cyclic compound
organic cation transport
synaptic vesicle exocytosis
protein processing
neuronal action potential
sensory perception of pain
cytolysis
protein catabolic process
positive regulation of bone mineralization
cellular response to extracellular stimulus
bleb assembly
positive regulation of prostaglandin secretion
response to lipopolysaccharide
positive regulation of interleukin-1 beta production
positive regulation of interleukin-6 production
collagen metabolic process
response to ATP
response to fluid shear stress
purinergic nucleotide receptor signaling pathway
T cell proliferation
T cell homeostasis
positive regulation of apoptotic process
positive regulation of catalytic activity
NAD transport
negative regulation of MAPK cascade
positive regulation of MAPK cascade
phospholipid translocation
positive regulation of ossification
negative regulation of bone resorption
positive regulation of cytolysis
ceramide biosynthetic process
pore complex assembly
skeletal system morphogenesis
homeostasis of number of cells within a tissue
positive regulation of protein secretion
defense response to Gram-positive bacterium
release of sequestered calcium ion into cytosol
protein complex oligomerization
positive regulation of cytoskeleton organization
response to calcium ion
response to electrical stimulus
membrane depolarization
positive regulation of mitochondrial depolarization
positive regulation of lymphocyte apoptotic process
cellular response to organic cyclic compound
reactive oxygen species metabolic process
apoptotic signaling pathway
extrinsic apoptotic signaling pathway
cation transmembrane transport
magnesium ion binding
lipopolysaccharide binding
purinergic nucleotide receptor activity
extracellularly ATP-gated cation channel activity
signaling receptor binding
copper ion binding
protein binding
ATP binding
zinc ion binding
channel activity
signaling receptor activity
protein serine/threonine kinase activator activity
scaffold protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P188222 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732414 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9227 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504956 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215793 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674920 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373586 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503931 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565757 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568140 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251318 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958460 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860204 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342827 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252442 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952727 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25898 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818246 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854996 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734113 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753266 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746811 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156765 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33173 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5572 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126964 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P188222 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P188222 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P188222 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P188222 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701458512 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 702132727 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 68591 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 65942 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 57501 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 702132727 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 701458512 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10762003 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10761996 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10761993 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762005 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762000 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762006 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762002 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10761995 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762004 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762001 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10761998 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10761994 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10761997 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10761999 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10761992 | - | - |
Rat Genome 230 2.0 Array | 1387718_PM_at | - | - |
Rat Genome 230 2.0 Array | 1391579_PM_at | - | - |
Rat Genome 230 2.0 Array | 1387718_at | - | - |
Rat Genome 230 2.0 Array | 1391579_at | - | - |
Affymetrix HT Rat Focus Array | 1387718_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1387718_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1200013508.rn.1 | TC1200001586.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200013509.rn.1 | TC1200001586.rn.1 | RTA TC |
Rat Expression Set 230 | 1387718_PM_at | - | - |
Rat Expression Set 230 | 1387718_at | - | - |
Rat Expression Set 230 | 1391579_PM_at | - | - |
Rat Expression Set 230 | 1391579_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10761992 | - | - |
Rat Genome U34 Set | X95882_at | - | - |
Rat Genome U34 Set | X95882_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22364257 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1370429 | - | - |
RatRef-12 Expression BeadChip | ILMN_59862 | - | - |
