Assay Details
Gene Symbol: P2rx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019256.1 | NP_062129.1 | 6 - 7 | 740 | 80 | |
XM_008769223.1 | XP_008767445.1 | 6 - 7 | 914 | 80 | ||
GenBank mRNA | FJ436445.1 | - | 6 - 7 | 778 | 80 | |
X95882.1 | - | 6 - 7 | 740 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
