Assay Details
Gene Symbol: Pptc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107141.1 | NP_001100611.1 | 5 - 6 | 1144 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P121881 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P121876 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816766 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265454 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303935 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785741 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255379 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380121 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669953 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705583 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87695 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897249 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790188 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382351 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661454 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750478 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798140 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698570 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195053 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157520 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37358 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157206 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109388 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P121876 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P121881 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P121876 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P121881 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P121881 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P121876 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P14713 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P102708 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 97434 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10762050 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762048 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762047 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762046 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762049 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762045 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10762044 | - | - |
Rat Genome 230 2.0 Array | 1378246_PM_at | - | - |
Rat Genome 230 2.0 Array | 1379953_at | - | - |
Rat Genome 230 2.0 Array | 1379953_PM_at | - | - |
Rat Genome 230 2.0 Array | 1388522_at | - | - |
Rat Genome 230 2.0 Array | 1378246_at | - | - |
Rat Genome 230 2.0 Array | 1388522_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1379953_at | - | - |
Affymetrix HT Rat Focus Array | 1379953_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1378246_at | - | - |
Affymetrix HT Rat Focus Array | 1388522_at | - | - |
Affymetrix HT Rat Focus Array | 1388522_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1378246_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1200013605.rn.1 | TC1200001596.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200013606.rn.1 | TC1200001596.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200013604.rn.1 | TC1200001596.rn.1 | RTA TC |
Rat Expression Set 230 | 1388522_at | - | - |
Rat Expression Set 230 | 1388522_PM_at | - | - |
Rat Expression Set 230 | 1379953_at | - | - |
Rat Expression Set 230 | 1379953_PM_at | - | - |
Rat Expression Set 230 | 1378246_at | - | - |
Rat Expression Set 230 | 1378246_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762044 | - | - |
Rat Genome U34 Set | rc_AI170820_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20819526 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369617 | - | - |
