Assay Details
Gene Symbol: Usp30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107153.1 | NP_001100623.1 | 12 - 13 | 1410 | 138 | |
XM_008769337.1 | XP_008767559.1 | 12 - 13 | 1415 | 138 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P530071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723055 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901239 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15708 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64538 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404297 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623124 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690359 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913029 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249262 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927176 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284839 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172575 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157960 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150935 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45858 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P530071 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P530071 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P530071 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P498990 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 30246 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 26046 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 37734 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10762858 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762865 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762863 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762862 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762859 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762860 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762869 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762867 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762861 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762866 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762870 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762868 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762864 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10762857 | - | - |
Rat Genome 230 2.0 Array | 1372570_at | - | - |
Rat Genome 230 2.0 Array | 1395360_PM_at | - | - |
Rat Genome 230 2.0 Array | 1395360_at | - | - |
Rat Genome 230 2.0 Array | 1372570_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1395360_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1372570_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1372570_at | - | - |
Affymetrix HT Rat Focus Array | 1395360_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1200015377.rn.1 | TC1200001787.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200015372.rn.1 | TC1200001787.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200015373.rn.1 | TC1200001787.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200015374.rn.1 | TC1200001787.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200015376.rn.1 | TC1200001787.rn.1 | RTA TC |
Rat Expression Set 230 | 1372570_PM_at | - | - |
Rat Expression Set 230 | 1372570_at | - | - |
Rat Expression Set 230 | 1395360_at | - | - |
Rat Expression Set 230 | 1395360_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762857 | - | - |
Rat Genome U34 Set | rc_AA996777_s_at | - | - |
Rat Genome U34 Set | rc_AI145892_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22117376 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355480 | - | - |
