Assay Details
Gene Symbol: Ogfod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105927.1 | NP_001099397.1 | 1 - 2 | 144 | 108 | |
XM_006249331.1 | XP_006249393.1 | 1 - 2 | 144 | 108 | ||
GenBank mRNA | BC167103.1 | - | 1 - 2 | 63 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
