Assay Details
Gene Symbol: Psph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009679.1 | NP_001009679.1 | 1 - 2 | 217 | 82 | |
GenBank mRNA | BC088310.1 | - | 1 - 2 | 217 | 82 | |
FQ213675.1 | - | 1 - 2 | 208 | 82 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC088310.1, NM_001009679.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
