Assay Details
Gene Symbol: Psph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009679.1 | NP_001009679.1 | 2 - 3 | 377 | 85 | |
GenBank mRNA | BC088310.1 | - | 2 - 3 | 377 | 85 | |
FQ213675.1 | - | 2 - 3 | 368 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
