Assay Details
Gene Symbol: Galnt9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107151.2 | NP_001100621.1 | 3 - 4 | 408 | 91 | |
NM_001122644.1 | NP_001116116.1 | 8 - 9 | 1720 | 91 | ||
XM_008769333.1 | XP_008767555.1 | 8 - 9 | 1697 | 91 | ||
XM_008769334.1 | XP_008767556.1 | 8 - 9 | 1719 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
