Assay Details
Gene Symbol: Galnt9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107151.2 | NP_001100621.1 | 5 - 6 | 672 | 102 | |
NM_001122644.1 | NP_001116116.1 | 10 - 11 | 1984 | 102 | ||
XM_008769333.1 | XP_008767555.1 | 10 - 11 | 1961 | 102 | ||
XM_008769334.1 | XP_008767556.1 | 10 - 11 | 1983 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P028953 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675876 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906676 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501241 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892636 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745741 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336040 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638839 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854348 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739288 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910919 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142017 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170367 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221977 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243968 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799789 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960804 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896302 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344725 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951268 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696388 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340657 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691504 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362693 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439393 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544422 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436839 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521994 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837256 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156540 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949982 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 551917 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784131 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647757 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905383 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582833 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289620 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908530 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146567 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958732 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881470 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260852 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64106 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360200 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541239 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89933 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3196 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52311 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77180 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174442 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 1257 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112459 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P028953 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P028953 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P028953 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P196625 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 62490 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 36922 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 72842 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10763007 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763009 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763011 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763008 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763013 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763001 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763005 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763004 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763003 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763006 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763010 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763012 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763002 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10763000 | - | - |
Affymetrix Rat Clariom S Assay | PSR1200015827.rn.1 | TC1200001862.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10763000 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21748337 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1356558 | - | - |
