Assay Details
Gene Symbol: Galnt9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122644.1 | NP_001116116.1 | 1 - 2 | 551 | 65 | |
XM_008769333.1 | XP_008767555.1 | 1 - 2 | 585 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
