Assay Details
Gene Symbol: Galnt9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122644.1 | NP_001116116.1 | 6 - 7 | 1394 | 56 | |
XM_008769333.1 | XP_008767555.1 | 6 - 7 | 1428 | 56 | ||
XM_008769334.1 | XP_008767556.1 | 6 - 7 | 1393 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
