Assay Details
Gene Symbol: Nsun5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191593.1 | NP_001178522.1 | 5 - 6 | 637 | 64 | |
XM_008769129.1 | XP_008767351.1 | 5 - 6 | 689 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
