Assay Details
Gene Symbol: Tfip11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008291.1 | NP_001008292.1 | 10 - 11 | 1874 | 116 | |
GenBank mRNA | BC085809.1 | - | 10 - 11 | 1874 | 116 | |
DQ342031.1 | - | 10 - 11 | 1873 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
