Assay Details
Gene Symbol: Cct6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033684.1 | NP_001028856.1 | 6 - 7 | 741 | 83 | |
GenBank mRNA | BC104703.1 | - | 6 - 7 | 741 | 83 | |
FQ216833.1 | - | 6 - 7 | 759 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
