Assay Details
Gene Symbol: Gnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031037.2 | NP_112299.1 | 1 - 2 | 143 | 140 | |
GenBank mRNA | BC065579.1 | - | 1 - 2 | 143 | 140 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC065579.1, NM_031037.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
