Assay Details
Gene Symbol: Acads | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022512.2 | NP_071957.1 | 8 - 9 | 1068 | 95 | |
GenBank mRNA | BC072545.1 | - | 8 - 9 | 1087 | 95 | |
FQ227562.1 | - | 2 - 3 | 239 | 95 | ||
J05030.1 | - | 8 - 9 | 1069 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
