Assay Details
Gene Symbol: Ankle2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047901.1 | NP_001041366.1 | 12 - 13 | 2581 | 117 | |
XM_008769342.1 | XP_008767564.1 | 11 - 12 | 5421 | 117 | ||
XM_008769343.1 | XP_008767565.1 | 11 - 12 | 5418 | 117 | ||
GenBank mRNA | AY325182.1 | - | 12 - 13 | 2581 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P524830 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894949 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414970 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79305 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138410 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14054 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208836 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235671 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293551 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136744 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765764 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17932 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829542 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495567 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726883 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35447 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355406 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266601 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424830 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742799 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145547 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99741 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48516 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32647 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P524830 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P524830 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P524830 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P243321 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P444759 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P524830 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 85069 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 50237 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 6634 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 72691 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10763086 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763089 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763083 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763076 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763087 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763079 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763081 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763077 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763080 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763090 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763078 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763074 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763088 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763085 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763084 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763082 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10763073 | - | - |
Rat Genome 230 2.0 Array | 1398522_at | - | - |
Rat Genome 230 2.0 Array | 1376003_PM_at | - | - |
Rat Genome 230 2.0 Array | 1376003_at | - | - |
Rat Genome 230 2.0 Array | 1398522_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1398522_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1376003_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1376003_at | - | - |
Affymetrix HT Rat Focus Array | 1398522_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1200015921.rn.1 | TC1200001871.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200015922.rn.1 | TC1200001871.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200015918.rn.1 | TC1200001871.rn.1 | RTA TC |
Rat Expression Set 230 | 1376003_at | - | - |
Rat Expression Set 230 | 1376003_PM_at | - | - |
Rat Expression Set 230 | 1398522_at | - | - |
Rat Expression Set 230 | 1398522_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10763073 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1367796 | - | - |
