Assay Details
Gene Symbol: Ankle2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047901.1 | NP_001041366.1 | 4 - 5 | 915 | 121 | |
XM_008769342.1 | XP_008767564.1 | 3 - 4 | 3755 | 121 | ||
XM_008769344.1 | XP_008767566.1 | 3 - 4 | 3755 | 121 | ||
GenBank mRNA | AY325182.1 | - | 4 - 5 | 915 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
