Assay Details
Gene Symbol: Ankle2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047901.1 | NP_001041366.1 | 5 - 6 | 1106 | 115 | |
XM_008769342.1 | XP_008767564.1 | 4 - 5 | 3946 | 115 | ||
XM_008769343.1 | XP_008767565.1 | 4 - 5 | 3943 | 115 | ||
XM_008769344.1 | XP_008767566.1 | 4 - 5 | 3946 | 115 | ||
GenBank mRNA | AY325182.1 | - | 5 - 6 | 1106 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
