Assay Details
Gene Symbol: Rfc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107146.2 | NP_001100616.1 | 1 - 2 | 138 | 104 | |
GenBank mRNA | BC168902.1 | - | 1 - 2 | 138 | 104 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC168902.1, NM_001107146.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
