Assay Details
Gene Symbol: Rfc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107146.2 | NP_001100616.1 | 9 - 10 | 950 | 71 | |
GenBank mRNA | BC168902.1 | - | 9 - 10 | 950 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
