Assay Details
Gene Symbol: Dtx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107157.1 | NP_001100627.1 | 5 - 6 | 1355 | 113 | |
XM_006249158.1 | XP_006249220.1 | 4 - 5 | 1301 | 113 | ||
XM_006249159.1 | XP_006249221.1 | 4 - 5 | 1301 | 113 | ||
XM_008769137.1 | XP_008767359.1 | 4 - 5 | 1156 | 113 | ||
GenBank mRNA | AY724523.1 | - | 3 - 4 | 1136 | 113 | |
BC166738.1 | - | 4 - 5 | 1242 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
