Assay Details
Gene Symbol: Dtx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107157.1 | NP_001100627.1 | 8 - 9 | 1669 | 86 | |
XM_006249158.1 | XP_006249220.1 | 7 - 8 | 1615 | 86 | ||
XM_006249159.1 | XP_006249221.1 | 6 - 7 | 1477 | 86 | ||
XM_008769137.1 | XP_008767359.1 | 7 - 8 | 1470 | 86 | ||
GenBank mRNA | AY724523.1 | - | 5 - 6 | 1312 | 86 | |
BC166738.1 | - | 7 - 8 | 1556 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
