Assay Details
Gene Symbol: Fbxw8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107145.1 | NP_001100615.1 | 3 - 4 | 588 | 100 | |
XM_008769233.1 | XP_008767455.1 | 3 - 4 | 615 | 100 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001107145.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
