Assay Details
Gene Symbol: Fbxw8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107145.1 | NP_001100615.1 | 6 - 7 | 1080 | 57 | |
XM_008769233.1 | XP_008767455.1 | 7 - 8 | 1265 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
