Assay Details
Gene Symbol: Ripk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139342.1 | NP_647558.1 | 5 - 6 | 73 | ||
XM_006251928.2 | XP_006251990.1 | 4 - 5 | 1775 | 73 | ||
XM_008770661.1 | XP_008768883.1 | 3 - 4 | 736 | 73 | ||
XM_008770662.1 | XP_008768884.1 | 4 - 5 | 1107 | 73 | ||
GenBank mRNA | AF036537.2 | - | 5 - 6 | 73 | ||
BC158580.1 | - | 3 - 4 | 490 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
