Assay Details
Gene Symbol: Ripk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139342.1 | NP_647558.1 | 9 - 10 | 1365 | 71 | |
XM_006251928.2 | XP_006251990.1 | 8 - 9 | 2217 | 71 | ||
XM_008770661.1 | XP_008768883.1 | 7 - 8 | 1178 | 71 | ||
XM_008770662.1 | XP_008768884.1 | 8 - 9 | 1549 | 71 | ||
GenBank mRNA | AF036537.2 | - | 9 - 10 | 1365 | 71 | |
BC158580.1 | - | 7 - 8 | 932 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
