Assay Details
Gene Symbol: Gsr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053906.2 | NP_446358.2 | 11 - 12 | 1000 | 129 | |
GenBank mRNA | CK357934.1 | - | 4 - 5 | 463 | 129 | |
U73174.1 | - | 1139 | 129 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
