Assay Details
Gene Symbol: Gsr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053906.2 | NP_446358.2 | 12 - 13 | 1140 | 65 | |
GenBank mRNA | CK357934.1 | - | 5 - 6 | 603 | 65 | |
U73174.1 | - | 1279 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
glutathione metabolic process
spermatogenesis
response to vitamin A
response to nicotine
cell redox homeostasis
response to ethanol
oxidation-reduction process
glutathione-disulfide reductase activity
protein homodimerization activity
glutathione binding
flavin adenine dinucleotide binding
NADP binding
glutathione metabolic process
spermatogenesis
response to vitamin A
response to nicotine
cell redox homeostasis
response to ethanol
oxidation-reduction process
glutathione-disulfide reductase activity
protein homodimerization activity
glutathione binding
flavin adenine dinucleotide binding
NADP binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P519868 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825198 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272628 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80527 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429679 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408734 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621834 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621256 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145745 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334354 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420765 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936004 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931858 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394462 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286815 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947196 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292374 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295403 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320072 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37581 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225101 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549893 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401256 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545296 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763487 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105447 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47257 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154122 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102838 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P519868 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P519868 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P519868 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P519868 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701243474 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 103175 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 52167 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 36294 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 701243474 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10792071 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10792065 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10792067 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10792075 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10792062 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10792066 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10792063 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10792064 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10792069 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10792068 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10792072 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10792061 | - | - |
Rat Genome 230 2.0 Array | 1369061_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369061_at | - | - |
Affymetrix HT Rat Focus Array | 1369061_at | - | - |
Affymetrix HT Rat Focus Array | 1369061_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1600013218.rn.1 | TC1600001797.rn.1 | RTA TC |
Rat Expression Set 230 | 1369061_at | - | - |
Rat Expression Set 230 | 1369061_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10792061 | - | - |
Rat Genome U34 Set | rc_AA893189_at | - | - |
Rat Genome U34 Set | U73174_at | - | - |
Rat Genome U34 Set | U73174_g_at | - | - |
Rat Genome U34 Set | AA851050_at | - | - |
Rat Genome U34 Set | U73174_g_at | - | - |
Rat Genome U34 Set | U73174_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21692763 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1352580 | - | - |
RatRef-12 Expression BeadChip | ILMN_58453 | - | - |
