Assay Details
Gene Symbol: Grtp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031813.2 | NP_001026983.1 | 3 - 4 | 401 | 108 | |
NM_001170477.1 | NP_001163948.1 | 4 - 5 | 563 | 108 | ||
GenBank mRNA | BC097985.1 | - | 3 - 4 | 355 | 7444444 | 108 |
CB315310.1 | - | 3 - 4 | 404 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
