Assay Details
Gene Symbol: Mak16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014002.1 | NP_001014024.1 | 1 - 2 | 57 | 99 | |
GenBank mRNA | BC090035.1 | - | 1 - 2 | 57 | 7367321 | 99 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC090035.1,NM_001014002.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
