Assay Details
Gene Symbol: Plat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013151.2 | NP_037283.2 | 1 - 2 | 85 | 62 | |
GenBank mRNA | BC061565.1 | - | 1 - 2 | 85 | 62 |
Gene Symbol: LOC100910418 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751623.3 | XP_003751671.2 | 1 - 2 | 89 | 62 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC061565.1, NM_013151.2, XM_003751623.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
